missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cholic Acid Sodium Salt, ≥97% (dry basis), Ultrapure, Affymetrix/USB™
Marca: Thermo Scientific J13630.A1
Detalles adicionales : Peso : 1.00000kg
Cantidad. | 1kg |
---|---|
Envase | Plastic bottle |
Identificadores de productos químicos
361-09-1 | |
430.561 | |
NRHMKIHPTBHXPF-RLWQFIAZSA-M | |
57349315 | |
CC(CCC(=O)[O-])C1CCC2C1(C(CC3C2C(CC4C3(CCC(C4)O)C)O)O)C.[Na+] |
C24H39NaO5 | |
MFCD00150749 | |
sodium cholate, sodium 3,7,12-trihydroxycholan-24-oate, sodium cholate, reagent, ksc575s0b, 3,7,12-trihydroxycholan-24-oic acid sodium salt | |
sodium;(4R)-4-[(8R,9S,10S,13R,14S,17R)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoate |
Especificaciones
Cholic Acid Sodium Salt | |
Dry basis | |
MFCD00150749 | |
NRHMKIHPTBHXPF-RLWQFIAZSA-M | |
sodium;(4R)-4-[(8R,9S,10S,13R,14S,17R)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoate | |
57349315 | |
Powder | |
Ultrapure | |
1kg |
361-09-1 | |
C24H39NaO5 | |
sodium cholate, sodium 3,7,12-trihydroxycholan-24-oate, sodium cholate, reagent, ksc575s0b, 3,7,12-trihydroxycholan-24-oic acid sodium salt | |
CC(CCC(=O)[O-])C1CCC2C1(C(CC3C2C(CC4C3(CCC(C4)O)C)O)O)C.[Na+] | |
430.561 | |
430.55 | |
≥98% | |
Plastic bottle |