missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloroform, ACS Reagent, contains ethanol as stabilizer, ≥99.8%, Honeywell Riedel-de Haën™
Click to view available options
Cantidad:
500 mL
1 L
2.5 L
15 L
20 L
4 x 2.5 L
Envase:
Frasco de vidrio
Glass Bottle
Stainless Steel Can
Descripción
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Especificaciones
Nombre del producto químico o material | Chloroform |
Nombre de nota | ACS Reagent, contains ethanol as stabilizer, ≥99.8% |
% mín. CAS | ≥90.0000%, ≥0.1000% |
% máx. CAS | ≤100.0000%, <1.0000% |
Punto de fusión | -63°C (literature) |
Densidad | 1.48 g/mL at 25 °C; 1.492 g/mL at 25 °C(lit.) |
Punto de ebullición | 60°C to 62°C |
Envase | Glass Bottle |
Fórmula lineal | [-CH2N(CH2CO2Na)CH2CO2H]2 |
Cantidad | 4 x 2.5 L |
Mostrar más |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'