missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlorobenzene, ≥99.5%, ACS Reagent, Honeywell Riedel-de Haën™
Click to view available options
Cantidad:
500 mL
1 L
2.5 L
Envase:
Glass Bottle
Descripción
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Especificaciones
Nombre del producto químico o material | Chlorobenzene |
Punto de fusión | -45°C |
Número UN | UN1134 |
Beilstein | 605632 |
Formula Weight (peso de la fórmula) | 112.56g/mol |
Porcentaje de pureza | ≥99.5% |
Grado | ACS Reagent |
Densidad | 1.106g/mL at 25°C |
Punto de ebullición | 132°C |
Cantidad | 500 mL |
Mostrar más |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'