Learn More
Chlormezanone 1.0 mg/ml in Methanol, LoGiCal
Product Code. 30462677
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Product Code. Quantity Unit Size
30462677 1 mL 1mL
1 options
This item is not returnable. View return policy
Product Code. 30462677 Supplier LoGiCal Supplier No. LGCAMP0017.0001
 Restricted Item
Fecha estimada de envoi: 22-04-2027
Add to basket
Add to basket
This item is not returnable. View return policy
LoGiCal delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
Chlormezanone 1.0 mg/ml in Methanol
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Chlormezanone
CAS 80-77-3
Analyte or Component Names Chlormezanone
Shipping Condition Ice Pack (-20°C)
Molecular Formula C11 H12 Cl N O3 S
Certifications/Compliance ISO 17034
Synonym Chlormezanone
InChI Formula InChI=1 S/C11H12ClNO3S/c1-13-10(14)6-7-17(15,16)11(13)8-2-4-9(12)5-3-8/h2-5,11 H,6-7H2,1H3
SMILES CN1C(c2ccc(Cl)cc2)S(=O)(=O)CCC1=O
IUPAC Name 2-(4-chlorophenyl)-3-methyl-1,1-dioxo-1,3-thiazinan-4-one
Solvent or Matrix Methanol
Industry Type Forensics
Molecular Weight (g/mol) 273.74
Quantity 1 mL
Formula Weight 273.0226 g/mol