Learn More
Chloramphenicol EP Impurity A, Mikromol™
Product Code. 17922726
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17922726 100 mg 100mg
30049075 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17922726 Supplier Mikromol Supplier No. MM0516.01
 Restricted Item
Fecha estimada de envoi: 26-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Chloramphenicol EP Impurity A
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (D)-(-)-threo-2-Amino-1-(4-nitrophenyl)-1,3-propanediol
CAS 716-61-0
Analyte or Component Names (D)-(-)-threo-2-Amino-1-(4-nitrophenyl)-1,3-propanediol
Shipping Condition Room Temperature
Molecular Formula C9 H12 N2 O4
Certifications/Compliance ISO 17025
Synonym (R,R)-(D)-(-)-threo-2-Amino-1-(4-nitrophenyl)-1,3-propanediol, 1,3-Propanediol, 2-amino-1-(4-nitrophenyl)-, (1R,2R)- (9CI), 1,3-Propanediol, 2-amino-1-(4-nitrophenyl)-, [R-(R*,R*)]-, 1,3-Propanediol, 2-amino-1-(p-nitrophenyl)-, D-threo-(-)- (8CI), (-)-threo-1-(p-Nitrophenyl)-2-amino-1,3-propanediol, (1R,2R)-(-)-2-Amino-1-(4-nitrophenyl)-1,3-propanediol, (1R,2R)-2-Amino-1-(4-nitrophenyl)-1,3-propanediol, (1R,2R)-threo-(-)-1-(4-Nitrophenyl)-2-amino-1,3-propanediol, Chloramphenicol D base, Chloramphenicol base, D-(-)-threo-1-p-Nitrophenyl-2-amino-1,3-propanediol, D-(-)-trans-1-p-Nitrophenyl-2-amino-1,3-propanediol, D-1-(p-Nitrophenyl)-2-aminopropane-1,3-diol, D-threo-(-)-2-Amino-1-(4-nitrophenyl)-1,3-propanediol, D-threo-(1R,2R)-1-p-Nitrophenyl-2-amino-1,3-propanediol, D-threo-2-Amino-1-(4-nitrophenyl)-1,3-propanediol, D-threo-2-Amino-1-(p-nitrophenyl)-1,3-propanediol, Levoamine, Chloramphenicol Impurity A
InChI Formula InChI=1S/C9H12N2O4/c10-8(5-12)9(13)6-1-3-7(4-2-6)11(14)15/h1-4,8-9,12-13H,5,10H2/t8-,9-/m1/s1
SMILES N[C@H](CO)[C@H](O)c1ccc(cc1)[N+](=O)[O-]
IUPAC Name (1R,2R)-2-amino-1-(4-nitrophenyl)propane-1,3-diol
Molecular Weight (g/mol) 212.2
Quantity 100 mg
Formula Weight 212.0797 g/mol