missing translation for 'onlineSavingsMsg'
Learn More

ChemR23/CMKLR1 Antibody (BZ194), Alexa Fluor™ 532, Novus Biologicals™

Rat Monoclonal Antibody

Marca:  Bio-Techne NBP1-43233AF532

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18216111

  • 503.23€ / 0.10 ml
Fecha estimada de envío: 21-08-2024
para ver el stock

Descripción

Descripción

ChemR23/CMKLR1 Monoclonal specifically detects ChemR23/CMKLR1 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.
Especificaciones

Especificaciones

ChemR23/CMKLR1
Monoclonal
Alexa Fluor 532
50 mM sodium borate with 0.05% sodium azide
CMKLR1
The immunogen for this antibody was CMKLR1.
Primary
Mouse
IgG2a κ
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
BZ194
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin
CHEMERINR, chemokine receptor-like 1, chemokine-like receptor 1, CHEMR23, DEZ, G-protein coupled receptor ChemR23, G-protein coupled receptor DEZ, MGC126105, MGC126106, orphan G-protein coupled receptor, Dez
Rat
0.1 mL
1240
Store at 4°C in the dark.
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales