Learn More
Faster™ ChemFAST Elite Recirculating Fume Hood
Product Code. 16258470
Change view
Click to view available options
For Use With (Equipment):
ChemFAST Elite 06
ChemFAST Elite 09
ChemFAST Elite 12
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
16258470 ChemFAST Elite 09 Each
16248470 ChemFAST Elite 06 Each
15938406 ChemFAST Elite 12 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16258470 Supplier Faster™ Supplier No. F00001840000
  • ONLINE EXCLUSIVE PRICE
    9069.01€
    8055.47€ / paquete de 1
    Ahorro 1013.54€
    11% Descuento
 Restricted Item
Fecha estimada de envoi: 22-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
ChemFAST ductless fume cupboards with innovative molecular filtration technology are made of the highest quality to guarantee a safe working environment with fume containment for protection from chemicals, vapors and aerosol.
Best cleanability: Front stratified hinged safety glass, the frontal opening is set to 200 mm. The whole front of the cabinet can also be opened upwards as it is hinged on the top – in order to allow easy access for complete and effective cleaning and decontamination.
Cold-rolled, epoxy painted, galvanized external structure for high resistance against chemical solvents aggression. Acid-resistant PVC rear wall with acid-resistant work surface.
High Level Lighting: The safety glass side-windows with the ideal positioning and sizing of the light-system provide the highest level of luminosity to the work area.
Easy handling and maintenance: ChemFAST can pass through standard 800 mm wide door openings thanks to the overall depth of just 760 mm. All service operations are available from the front of the cabinet. Charcoal filters can be easily replaced from the front.
More than 10 different kind of activate charcoal filters able to absorb and handle any organic compounds and inorganic gases and vapors.
Highly effective protection from toxic vapors
High filtration efficiency
Large adsorption capacity
Built in safety: all electronic components, switches, lighting and motor fan have been selected and installed fully isolated from air contaminated with solvents and therefore meet the most stringent electrical safety requirements. Visual and audible alarms for prefilter clogging, filter saturation, anenometer failure, motor fan failure, and gas detection.
Specifications
Material Epoxy-coated Steel
Depth (English) 29.92 in.
Depth (Metric) 760 mm
Dimensions (L x W x H) 760 x 885 x 1120 mm
Flow Rate 400 m3/hr.
For Use With (Equipment) ChemFAST Elite 09
Includes Removable PVC Spillage Tray, Microprocessor System, Lighting, Prefilter
Location Bench, Stand
Width (English) 34.84 in.
Width (Metric) 885 mm