Learn More
CHEMetrics CHEMets Phenol Test Kit Refill
Product Code. 11771044
Change view
Click to view available options
No. of Tests:
30
Unit Size:
Pack of 30
Product Code. No. of Tests Unit Size
11771044 30 Pack of 30
1 options
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 11771044 Supplier CHEMetrics Supplier No. R8012
 Restricted Item
En stock
Add to basket
Edge
Add to basket
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Provides results in two minutes or less
This product contains 30 self-filling CHEMets ampules to refill kit K-8012 for the analysis of phenols. R-8012 is compatible with C-8001 and C-8012 comparators.
Specifications
Analytes Phenol
For Use With (Equipment) C-8001 and C-8012 comparators
Includes 30 self-filling CHEMets ampoules to refill kit K-8012
No. of Tests 30
Range 0 to 1 ppm and 1 to 12 ppm
Test Method 4-Aminoantipyrine
Type CHEMets Refill Kit
compliance-icons compliance-icons
Product Identifier
  • phenol vacu-vials, chemets, and vacuettes ampoules
Signal Word
  • Warning
Hazard Category
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H341-Suspected of causing genetic defects.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P201-Obtain special instructions before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P271-Use only outdoors or in a well-ventilated area.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: potassium ferricyanide(III)