missing translation for 'onlineSavingsMsg'
Learn More

CHCHD7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 19911-1-AP-20UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16819384

  • 109.00€ / 20 microlitros

Envío estimado: 02-11-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

CHCHD7 is a newly identified member of a multifamily of proteins containing a conserved (coiled coil 1)-(helix 1)-(coiled coil 2)-(helix 2) domain. Northern blot analysis revealed that the gene is ubiquitously expressed. Its biological function is unknown and the gene has hitherto not been associated with neoplasia. CHCHD7 and PLAG1 are located head-to-head about 500 bp apart in 8q12. A chromosomal aberration involving CHCHD7 is found in salivary gland pleiomorphic adenomas, the most common benign epithelial tumors of the salivary gland.
Especificaciones

Especificaciones

CHCHD7
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8K2Q5, Q9BUK0
CHCHD7
Rabbit
IgG
20 μL
-20°C
Primary
66433, 684258, 79145
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
CHCHD7
Liquid
Chchd7
CHCHD7 Fusion Protein Ag13735
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Rat, Mouse, Human
Videos
SDS
Documentos
Certificados