Learn More
Spectrum Labs™ CELLMAX IMPLANT MEMBRANES Spectrum
Product Code. 12883215
Change view
Click to view available options
Quantity:
3 Fibers/Pk.
Unit Size:
Pack of 2
Product Code. Quantity Unit Size
12883215 3 Fibers/Pk. Pack of 2
1 options
This item is not returnable. View return policy
Product Code. 12883215 Supplier Spectrum Labs™ Supplier No. M138615
  • ONLINE EXCLUSIVE PRICE
    152.00€
    130.88€ / paquete de 2
    Ahorro 21.12€
    13% Descuento
Fecha estimada de envoi: 28-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
Description Hollow fiber membrane for implant technology
For Use With (Application) Drug screening and cancer research
Material Modified Polyvinylidene Difluoride (mPVDF)
Quantity 3 Fibers/Pk.
Test Time 10 days