Learn More
Thermo Scientific™ CellInsight™ CX7 High Content Analysis Platform
Integrated system for high content analysis
en la oferta - 174800.00€
Alertas:
For research use only
Productos 2
Código de producto Marca Descripción Precio Cantidad y disponibilidad  
Código de producto Marca Descripción Precio Cantidad y disponibilidad  
15900603 Ver documentos Thermo Scientific™
CX7A1110
CellInsight™ CX7 High Content Analysis Platform en la oferta
cada uno Ver productos alternativos
Ahorre hasta 
»
N/A
Este ítem no se puede comprar online porque su precio está sujeto a variación. Por favor, contacte con nuestro equipo de soporte de producto en el 902 239 303 para recibir ayuda.
15910603 Ver documentos Thermo Scientific™
CX7B1112
CellInsight™ CX7 High Content Analysis Platform and Store Standard Edition 174800.00€
cada uno Ver productos alternativos
Ahorre hasta 
»
Fecha de envío estimada
del proveedor 24-10-2019
Añadir a la cesta
Descripción
Descripción
The Thermo Scientific™ CellInsight™ CX7 High Content Analysis (HCA) Platform is a fast, automated cellular imaging and analysis platform designed for quantitative microscopy.
The CellInsight CX7 HCA Platform offers an integrated way to develop and automate high content assays using a combination of 5-color brightfield and 7-color fluorescence imaging in confocal or wide-field modes. See how the CellInsight CX7 platform compares to other high content analysis platforms from Thermo Fisher Scientific.
Features of the CellInsight CX7 HCA Platform
  • 7-channel fluorescent imaging
  • 5-channel bright-field imaging
  • 7-channel confocal imaging
  • Software and laser-based autofocus for consistent scan times
  • Configured for fully automated plate handling and scanning
Capabilities
  • Addresses hundreds of assay types with flexible software and hardware tools
  • Scan and analyze microscope slides or most SBS-standard format microplates
  • Use HCS Studio software for integrated instrument control and data analysis
  • Achieve the fastest time to results by using intelligent acQuisition (iQ) real-time analysis feedback
  • Compatible with Molecular Probes™ fluorescent detection reagents
Hardware and Software
  • Stable and reliable light source enables reduced variability, increased data quality, and decreased operating costs
  • Broad reagent options with seven-color excitation from violet to near IR for assay multiplexing
  • Dual mode focusing using Software and Laser Autofocus methods to address both labelled and unlabeled sample types
  • Multi-mode imaging allows for wide-field, bright-field, and confocal imaging in one run
  • HCS Studio Cell Analysis Software is an integrated instrument control, visualization, and analysis software package for novice to advanced users
  • Scalable with Store Image and Database Software and optional Orbitor RS Microplate handler