Learn More
Cefixime Trihydrate, LoGiCal
Product Code. 30461549
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Product Code. Quantity Unit Size
30461549 10 mg 10mg
1 options
This item is not returnable. View return policy
Product Code. 30461549 Supplier LoGiCal Supplier No. LGCFOR0316.00
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This item is not returnable. View return policy
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Cefixime Trihydrate
CAS 125110-14-7
Analyte or Component Names Cefixime Trihydrate
Shipping Condition Room Temperature
Molecular Formula C16 H15 N5 O7 S2 . 3 H2 O
Certifications/Compliance ISO 9001
Synonym Cefixime Trihydrate, Cefixime, (6R,7R)-7-[[(Z)-2-(2-Aminothiazol-4-yl)-2-[(carboxymethoxy)imino]acetyl]amino]-3-ethenyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid trihydrate
InChI Formula InChI=1S/C16H15N5O7S2.3H2O/c1-2-6-4-29-14-10(13(25)21(14)11(6)15(26)27)19-12(24)9(20-28-3-8(22)23)7-5-30-16(17)18-7;;;/h2,5,10,14H,1,3-4H2,(H2,17,18)(H,19,24)(H,22,23)(H,26,27);3*1H2/b20-9-;;;/t10-,14-;;;/m1.../s1
Solution Type Neat
SMILES O.O.O.Nc1nc(cs1)\C(=N\OCC(=O)O)\C(=O)N[C@H]2[C@H]3SCC(=C(N3C2=O)C(=O)O)C=C
IUPAC Name (6R,7R)-7-[[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-(carboxymethoxyimino)acetyl]amino]-3-ethenyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;trihydrate
Industry Type Clinical Testing
Molecular Weight (g/mol) 507.5
Quantity 10 mg
Formula Weight 507.073 g/mol