Learn More
CEACAM5/CD66e Antibody (CB30), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Marca:  Novus Biologicals NB110-58734FR
Detalles adicionales : Peso : 0.00970kg
Código de producto. 18214060
  • 497.83€ / 0.10 ml
Fecha estimada de envío: 19-08-2024
para ver el stock
Descripción
Descripción
CEACAM5/CD66e Monoclonal specifically detects CEACAM5/CD66e in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Especificaciones
Especificaciones
CEACAM5/CD66e
Monoclonal
DyLight 680
Carcinoembryonic antigen, carcinoembryonic antigen-related cell adhesion molecule 5, CD66e antigen, CEACD66e, DKFZp781M2392, Meconium antigen 100
Mouse
Protein A purified
RUO
Primary
The antibody CB30 recognizes CD66e (CEA; 180-200 kDa), a cell surface bound carcinoembryonic antigen mainly expressed on epithelial cells.
Store at 4C in the dark.
IgG1
Flow Cytometry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
CB30
Flow Cytometry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
CEACAM5
Human carcinoembryonic antigen (CEA; CEACAM5)
0.1 mL
Cancer, Cellular Markers, Immunology
1048
Human
Purified