missing translation for 'onlineSavingsMsg'
Learn More

CEACAM1/CD66a Antibody (F7), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody

Marca:  Bio-Techne NBP2-59673PCP

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18214163

  • 564.78€ / 0.10 ml
Fecha estimada de envío: 21-08-2024
para ver el stock

Descripción

Descripción

CEACAM1/CD66a Monoclonal specifically detects CEACAM1/CD66a in Rat samples. It is validated for Flow Cytometry.
Especificaciones

Especificaciones

CEACAM1/CD66a
Monoclonal
PerCP
PBS with 0.05% Sodium Azide
CEACAM1
The immunogen for this antibody is proprietary.
0.1 mL
Apoptosis, Cancer
634
Rat
Purified
Flow Cytometry
F7
Flow Cytometry
antigen CD66, BGP-1, BGP1BGPBiliary glycoprotein 1, BGPI, biliary glycoprotein adhesion molecule, carcinoembryonic antigen-related cell adhesion molecule 1, carcinoembryonic antigen-related cell adhesion molecule 1 (biliaryglycoprotein), CD66a, CD66a antigen
Mouse
Protein A purified
RUO
Primary
Binds specific to the N-domain of rat CEACAM1. Does not cross-react with human CEACAM-1, -3. -4, -5, -6, 7 and -8, mouse CEACAM1 and mouse CEACAM2.
Store at 4C in the dark.
IgG1 κ
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales

For Research Use Only (RUO)