missing translation for 'onlineSavingsMsg'
Learn More

CDR2L Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 14563-1-AP-150UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16813264

  • 319.00€ / 150 microlitros

Envío estimado: 02-11-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

14563-1-AP recognizes a 62 kDa band due to phosphorylation.

Cerebellar degeneration-related protein 2-like (CDR2L), also named as HUMPPA or Paraneoplastic 62 kDa antigen, is a 465 amino acid protein, which belongs to the CDR2 family. CDR2L contains three potential coiled-coil regions. The functions of protein is so far not known. CDR2L is expressed in the brain and localizes in the cytoplasm. The predicted MW of this protein is 53 kDa.
Especificaciones

Especificaciones

CDR2L
0.2 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
A2A6T1, Q86X02
CDR2L, HUMPPA, Paraneoplastic 62 kDa antigen
Rabbit
IgG
150 μL
-20°C
Primary
237988, 30850, 360656
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
CDR2L
Liquid
Cdr2l
CDR2L Fusion Protein Ag6127
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Videos
SDS
Documentos
Certificados