missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human CDKL5, GST Tag Recombinant Protein

This CDKL5 Recombinant Human Protein (1-498) was expressed in insect cells.

Marca:  Invitrogen™ A33353

Detalles adicionales : Peso : 0.00900kg

 Ver más versiones de este producto

Código de producto. 17118775

  • 7580.00€ / 1 miligramo
Fecha estimada de envío: 30-04-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

CDKL5 serine/threonine kinase (STK9) is associated with X-linked infantile spasm syndrome (ISSX), also known as X-linked West syndrome and Rett syndrome (RTT). CDKL5 can control nuclear speckle morphology by regulating the phosphorylation state of splicing regulatory proteins, and may be involved indirectly in pre-mRNA processing by controlling splicing factor dynamics. The mutations in CDKL5 are a major cause of infantile spasms and early epileptic seizures in female patients, and of a later intractable seizure disorder.
  • Validated for use with the Z’-LYTE™ assay
  • Available in multiple pack sizes for convenient use
Especificaciones

Especificaciones

NP_003150
82.7 kDa
CDKs and CHKs
Human
GST Tag
Dry Ice
Recombinant, Active, Catalytic
Kinase Assay
1 mg
aa 1 - 498
Recombinant
Storage Buffer: 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol.
Insect
Liquid
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales