missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ CDK2 Recombinant Protein Antigen

Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition

Marca:  Novus Biologicals™ NBP2-57327PEP

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18215553

  • 242.97€ / 100 microlitros
Fecha estimada de envío: 14-08-2024
para ver el stock

Descripción

Descripción

A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human CDK2. Source: E.coli Amino Acid Sequence: PSFPKWARQDFSKVVPPLDEDGRSLLSQMLHYDPNKRIS The CDK2 Recombinant Protein Antigen is derived from E. coli. The CDK2 Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.
Especificaciones

Especificaciones

1017
CDK2 Recombinant Protein Antigen
PBS and 1M Urea, pH 7.4.
cdc2-related protein kinase, cell devision kinase 2, Cell division protein kinase 2, cyclin-dependent kinase 2, EC 2.7.11, EC 2.7.11.22, p33 protein kinase, p33(CDK2)
Unlabeled
100μL
E.coli
>80% by SDS-PAGE and Coomassie blue staining
Store at −20°C. Avoid freeze-thaw cycles.
Blocking/Neutralizing, Control
CDK2
Recombinant Protein Antigen
RUO
This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP2-52980. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/ml
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales

For Research Use Only.