Learn More
CD98 Antibody (071), Alexa Fluor™ 405, Novus Biologicals™
Rabbit Monoclonal Antibody
Marca:  Novus Biologicals NBP2-90076AF405
Detalles adicionales : Peso : 0.00970kg
Código de producto. 18213668
  • 607.98€ / 0.10 ml
Fecha estimada de envío: 14-08-2024
para ver el stock
Descripción
Descripción
CD98 Monoclonal specifically detects CD98 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Especificaciones
Especificaciones
CD98
Monoclonal
Alexa Fluor 405
50mM Sodium Borate
Rabbit
Protein A purified
RUO
Primary
Human
Purified
Immunohistochemistry, Immunohistochemistry (Paraffin)
71
Immunohistochemistry, Immunohistochemistry-Paraffin
4F2, 4F2hc, 4T2HC, antigen identified by monoclonal antibodies 4F2, TRA1.10, TROP4, and T43,4F2HC, CD98, CD98 antigen, CD98 heavy chain, CD98HC, heavy chain, Lymphocyte activation antigen 4F2 large subunit, MDU1antigen defined by monoclonal 4F2, heavy chain, monoclonal 44D7, NACAE4F2 cell-surface antigen heavy chain, solute carrier family 3 (activators of dibasic and neutral amino acidtransport), member 2,4F2 heavy chain antigen
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD98 (P08195-1; Arg206-Ala630).
0.1 mL
Biologically Active Proteins, Cell Biology, Cellular Markers, Immunology, Lipid and Metabolism, Plasma Membrane Markers, Signal Transduction
6520
Store at 4°C in the dark.