missing translation for 'onlineSavingsMsg'
Learn More

FAS Mouse anti-Human, Pacific Blue™, Clone: DX2, Invitrogen™

Mouse Monoclonal Antibody

Marca:  Invitrogen MHCD9528

Detalles adicionales : Peso : 0.50000kg

 Ver más versiones de este producto

Código de producto. 10797513

  • 349.00€ / cada uno


Fecha de envío estimada
del proveedor 05-12-2019
Añadir a la cesta

Descripción

Descripción

Fas ligand (FasL, CD 95L) is a type-II-membrane protein, whose N-terminus is in the cytoplasm and its C-terminal region extends into the extracellular space. Its receptor Fas (CD 95, Apo-1) is a cell-surface-type-I-membrane protein and a member of the tumor necrosis factor (TNF) and nerve growth factor (NGF) receptor family. As a member of the TNF-cytokine family FasL induces apoptosis when interacting with its receptor Fas. FasL may exist as either membrane bound (45 kD) or soluble forms (26 kD). The soluble protein can be released from cells upon cleavage by metalloproteinases. Binding of FasL to Fas leads to oligomerization of the receptor and triggers apoptotic cell death through the interaction of other proteins. FasL is predomitly expressed in activated T-lymphocytes and natural killer (NK) cells also it is expressed in the tissues of immune-privilege sites such as testis and eye. FasL expression is also reported in various tissues as thymus, liver, ovary, lung, heart and kidney. The Fas/FasL system has been shown to play a role in a number of human diseases, for example AIDS, hepatitis or cancer. It is assumed that induction of apoptosis through FasL is predomitly involved in anti-viral immune responses.
Especificaciones

Especificaciones

FAS
Monoclonal
Pacific Blue™
FAS
Liquid
FAS
Human CD95.
Purified
RUO
Antibody
Monoclonal
Human
Flow Cytometry
DX2
PBS with BSA and 0.1% sodium azide
P25445
ALPS1A, APO-1, APT1, Apo-1 Fas, CD95, FAS1, FASTM, TNFRSF6
Mouse
IgG1
500 μL
4° C
Primary
355
Videos
Documentos