missing translation for 'onlineSavingsMsg'
Learn More

CD90.2 (Thy-1.2) Rat anti-Mouse, Super Bright 645, Clone: 53-2.1, eBioscience™

Rat Monoclonal Antibody

Marca:  Invitrogen 64-0902-82

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15913634

  • 302.00€ / 100 microgramos


Fecha de envío estimada
del proveedor 03-05-2019
Añadir a la cesta

Descripción

Descripción

Light sensitivity: This tandem dye is sensitive to photo- induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (Product No. 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (Product No. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 405 nm; Emission: 645 nm; Laser: Violet Laser CD90 (Thy 1) antigen is a GPI linked glycoprotein member of the Immunoglobulin super family. It is expressed in murine T cells, thymocytes, neural cells, Kupffer′s cells and fibroblasts. Thy 1 may play a role in cell-cell or cell-ligand interactions during synaptogenesis and other events in the brain.
Especificación

Especificación

CD90.2 (Thy-1.2)
Monoclonal
0.2 mg/mL
PBS with BSA and 0.09% sodium azide; pH 7.2
P01831
Thy1
IgG2a, kappa
100 μg
4° C, store in dark, DO NOT FREEZE!
Primary
21838
Flow Cytometry
53-2.1
Super Bright 645
Thy1
Liquid
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse
Videos