missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD8a Rat anti-Mouse, BB700, Clone: 53-6.7, BD Horizon™
Rat Monoclonal Antibody
Marca: BD Biosciences 566410
Detalles adicionales : Peso : 0.00900kg
Descripción
CD8a Monoclonal antibody specifically detects CD8a in Mouse samples. It is validated for Flow Cytometry.Especificaciones
CD8a | |
Monoclonal | |
0.2 mg/mL | |
Rat | |
RUO | |
Mouse | |
Purified |
Flow Cytometry | |
53-6.7 | |
Brilliant Blue 700 (BB700) | |
25 μg | |
Primary | |
Refrigerate (2 to 8°C) | |
IgG2a κ |