missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD86 Rat, FITC, Clone: GL-1 (GL1), enQuireBio™
Rat Monoclonal Antibody
Marca: EnquireBio QAB51-F-25ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
CD86 | |
Monoclonal | |
0.5mg/mL | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
Cd86 | |
Protein A/G Purified | |
RUO | |
12524 | |
IgG2a κ |
Flow Cytometry | |
GL-1 (GL1) | |
FITC | |
NP_062261 | |
Rat | |
25 μg | |
Primary | |
Store at 2-4°C. |