missing translation for 'onlineSavingsMsg'
Learn More

CD81 / TAPA-1 Mouse, Clone: 1.3.3.22, enQuireBio™

Mouse Monoclonal Antibody

Marca:  EnquireBio 975-MSM1-P2

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15931308

  • 494.00€ / 200 microgramos


Fecha estimada de envío
14-10-2019
Añadir a la cesta

Especificaciones

Especificaciones

CD81 / TAPA-1
Cell surface
200μg/mL
10mM PBS with 0.05% BSA and 0.05% Azide
CD81
B-Cell line derived from a Burkitt lymphoma
26kDa
Protein A/G Purified
RUO
Primary
975
Immunohistochemistry
1.3.3.22
Unconjugated
NP_001284578
Mouse
IgG1 kappa
Ramos, Daudi, Raji, U266, HUT-78, HUT-102, Jurkat, HL-60, KG1a, A293, A431, MG63, and human lymphocytes. Tonsil or Liver.
200 μg
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
Monoclonal
Videos
Documentos