missing translation for 'onlineSavingsMsg'
Learn More

CD81 Antibody (2F7), PE, Novus Biologicals™

Armenian Hamster Monoclonal Antibody

Marca:  Bio-Techne NBP1-28136PE

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18214162

  • 502.15€ / 0.25 ml
Fecha estimada de envío: 19-08-2024
para ver el stock

Description

Description

CD81 Monoclonal specifically detects CD81 in Human, Mouse, Hamster samples. It is validated for Flow Cytometry.
Specifications

Specifications

CD81
Monoclonal
PE
PBS with 0.05% Sodium Azide
CD81
Mouse epithelial cell line PAM212
0.25 mL
Adaptive Immunity, Cell Cycle and Replication, Immunology
975
Human, Mouse, Hamster
Purified
Flow Cytometry
2F7
Flow Cytometry
CD81 antigen, CD81 antigen (target of antiproliferative 1), CD81 molecule, CVID6, TAPA1, TAPA-1, Target of the antiproliferative 1, Tetraspanin-28, Tspan-28, tspan-28,26 kDa cell surface protein TAPA-1, TSPAN28S5.7
Armenian Hamster
Protein A or G purified
RUO
Primary
Mouse CD81 (Mr 25 kDa), a member of the transmembrane 4 integral membrane protein family
Store at 4C in the dark.
IgG3 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Special Offers

Special Offers