missing translation for 'onlineSavingsMsg'
Learn More

CD80 (B7-1) Armenian Hamster anti-Canine, Mouse, Porcine, Super Bright 436, Clone: 16-10A1, eBioscience™

Armenian Hamster Monoclonal Antibody

Marca:  Invitrogen 62-0801-82

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15923394

  • 278.00€ / 100 microgramos


Fecha estimada de envío
20-08-2020
Añadir a la cesta

Descripción

Descripción

SB-4401) to minimize any non-specific polymer interactions. Please refer to the datasheet for Super Bright Staining Buffer for more information. Excitation: 405 nm; Emission: 436 nm; Laser: Violet Laser Super Bright Polymer Dyes are sold under license from Becton, Dickinson and Company. CD80 (B7-1) and CD86 (B7-2) are ligands of T cell critical costimulatory molecule CD28 and of an inhibitory receptor CTLA-4 (CD152). The both B7 molecules are expressed on professional antigen-presenting cells and are essential for T cell activation, the both molecules can also substitute for each other in this process. The question what are the differences in CD80 and CD86 competency has not been fully elucidated yet; there are still conflicts in results about their respective roles in initiation or sustaining of the T cell immune response.
Especificaciones

Especificaciones

CD80 (B7-1)
Monoclonal
0.2 mg/mL
PBS with BSA and 0.09% sodium azide; pH 7.2
Q00609
CD80
IgG
100 μg
4° C, store in dark, DO NOT FREEZE!
Primary
12519, 397161, 403765
Flow Cytometry
16-10A1
Super Bright 436
Cd80
Liquid
Armenian Hamster
Affinity chromatography
RUO
Antibody
Monoclonal
Canine, Mouse, Porcine
Videos
Documentos