missing translation for 'onlineSavingsMsg'
Learn More

CD80 Mouse anti-Human, Alexa Fluor™ 647, Clone: MEM-233, Invitrogen™

Mouse Monoclonal Antibody

Marca:  Invitrogen A51018

Detalles adicionales : Peso : 0.00460kg

Código de producto. 15692155

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
Ver productos alternativos »


Descripción

Descripción

CD80 (B7-1) and CD86 (B7-2) are ligands of T cell critical costimulatory molecule CD28 and of an inhibitory receptor CTLA-4 (CD152). The both B7 molecules are expressed on professional antigen-presenting cells and are essential for T cell activation, the both molecules can also substitute for each other in this process. The question what are the differences in CD80 and CD86 competency has not been fully elucidated yet; there are still conflicts in results about their respective roles in initiation or sustaining of the T cell immune response.
Especificaciones

Especificaciones

CD80
Monoclonal
0.12 mg/mL
PBS with 1% BSA and 0.0325% sodium azide; pH 7.4
P33681
CD80
Extracellular domain of human CD80 fused to human IgG1(Fc)
100 Tests
Primary
Human
Antibody
IgG1
Flow Cytometry
MEM-233
Alexa Fluor 647
CD80
B7-1, B7.1, Cd28l, Ly-53, Ly53, MIC17, TS/A-1, LAB7, B7.1, CD28LG, CD28LG1
Mouse
Affinity chromatography
RUO
941
4° C, store in dark
Liquid
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales