missing translation for 'onlineSavingsMsg'
Learn More

CD74 Antibody (CerCLIP.1), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody

Marca:  Novus Biologicals NBP1-19249AF750

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18213511

  • 465.43€ / 0.10 ml
Fecha estimada de envío: 21-08-2024
para ver el stock

Descripción

Descripción

CD74 Monoclonal specifically detects CD74 in Human samples. It is validated for Flow Cytometry.
Especificaciones

Especificaciones

CD74
Monoclonal
Alexa Fluor 750
50mM Sodium Borate with 0.05% Sodium Azide
CD74
Synthetic peptide derived from the sequence of human CLIP
0.1 mL
Immunology
Human
IgG1
Flow Cytometry
CerCLIP.1
Flow Cytometry
CD74 antigen, CD74 antigen (invariant polypeptide of major histocompatibility complex, classII antigen-associated), CD74 molecule, major histocompatibility complex, class II invariant chain, DHLAGgamma chain of class II antigens, HLA class II histocompatibility antigen gamma chain, HLADG, HLA-DR antigens-associated invariant chain, HLA-DR-gamma, Ia antigen-associated invariant chain, Ia-associated invariant chain, Ia-GAMMA, Ii, MHC HLA-DR gamma chain, p33
Mouse
Protein G purified
RUO
Primary
Store at 4°C in the dark.
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales