missing translation for 'onlineSavingsMsg'
Learn More

CD7 Mouse anti-Human, PE, Clone: CD7-6B7, Invitrogen™

Mouse Monoclonal Antibody

Marca:  Invitrogen MHCD0704

Detalles adicionales : Peso : 0.50000kg

Código de producto. 10445093

  • 260.00€ / cada uno


Fecha de envío estimada
del proveedor 25-02-2019
Añadir a la cesta

Descripción

Descripción

CD7 (gp40, Leu9) is a 40 kDa member of the immunoglobulin gene superfamily. CD7 contains N-terminal amino acids 1-107 are highly homologous to Ig kappa-L chains whereas the carboxy-terminal region of the extracellular domain is proline-rich and has been postulated to form a stalk from which the Ig domain projects. CD7 is expressed on the majority of immature and mature T-lymphocytes, and T cell leukemia. Further, CD7 is also found on natural killer cells, a small subpopulation of normal B cells and on maligt B cells. Cross-linking surface CD7 positively modulates T cell and NK cell activity as measured by calcium fluxes, expression of adhesion molecules, cytokine secretion and proliferation. CD7 associates directly with phosphoinositol 3′-kinase. CD7 ligation induces production of D-3 phosphoinositides and tyrosine phosphorylation. Expression of CD7 is an important marker used in leukemia diagnostics.
Especificación

Especificación

CD7, CD7
Monoclonal
PE
CD7
Liquid
CD7
Human CD7
Purified
ASR
Antibody
Monoclonal
Human
Flow Cytometry
CD7-6B7
PBS with 4mg/mL BSA, sucrose and 0.1% sodium azide
P09564
GP40, LEU-9, TP41, Tp40
Mouse
IgG2a
0.5 mL
4° C, store in dark
Primary
924
Videos