missing translation for 'onlineSavingsMsg'
Learn More

CD69 Monoclonal Antibody (FN50), Super Bright™ 600, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody

en la oferta

Especificaciones

Antígeno CD69
Clon FN50
Concentración 5 μL/Test
Aplicaciones Flow Cytometry
Clasificación Monoclonal
Ver más especificaciones

Descripción

Descripción

Description: The FN50 monoclonal antibody reacts with human CD69, also known as very early activation antigen (VEA). CD69 is approximately 30 kDa and is expressed on the cell-surface as a disulfide-linked dimer. CD69 is rapidly upregulated upon activation and expressed on lymphocytes, monocytes and platelets.

Applications Reported: This FN50 antibody has been reported for use in flow cytometric analysis. Applications Tested: This FN50 antibody has been pre-diluted and tested by flow cytometric analysis of normal human peripheral blood cells. This may be used at 5 μL (0.06 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Super Bright 600 is a tandem dye that can be excited with the violet laser line (405 nm) and emits at 600 nm. We recommend using a 610/20 bandpass filter. Please make sure that your instrument is capable of detecting this fluorochrome. When using two or more Super Bright dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Complete Staining Buffer (Product # SB-4401) to minimize any non-specific polymer interactions. Please refer to the datasheet for Super Bright Staining Buffer for more information. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (Product # 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (Product # 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 405 nm; Emission: 600 nm; Laser: Violet Laser Super Bright Polymer Dyes are sold under license from Becton, Dickinson and Company.
Especificaciones

Especificaciones

CD69
5 μL/Test
Monoclonal
Liquid
RUO
Q07108
CD69
Primary
4° C, store in dark, DO NOT FREEZE!
CD69
FN50
Flow Cytometry
Super Bright 600
Mouse
Human
969
IgG1 κ
Affinity chromatography
Antibody
Videos
SDS
Documentos

Documentos

Product Certifications