missing translation for 'onlineSavingsMsg'
Learn More

CD69 Mouse anti-Human, APC-eFluor® 780, Clone: FN50, eBioscience™

Mouse Monoclonal Antibody

Marca:  Invitrogen 47-0699-41

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15933294

  • 128.00€ / paquete de 25


Fecha de envío estimada
del proveedor 03-03-2020
Añadir a la cesta

Descripción

Descripción

Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 633-647 nm; Emission: 780 nm; Laser: Red Laser CD69 (AIM, Active Inducer Molecule) is a gp28/34 disulfide bonded homodimer with a molecular weight of 60 kDa under non-reducing conditions. CD69 contains one or two N linked oligosacaride and the molecule is present on activated platelets. In normal peripheral blood a variable percentage of cells express the CD69 antigen, and it is involved in lymphocyte signal transduction. Expression CD69 is induced upon activation of T lymphocytes, and may play a role in proliferation. Furthermore, the protein may act to transmit signals in natural killer cells and platelets. Alternative splicing results in multiple transcript variants of CD69. Diseases associated with CD69 dysfunction include coccidiodomycosis and asthma.
Especificaciones

Especificaciones

CD69
Monoclonal
5 μL/Test
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2
Q07108
CD69
IgG1, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
969
Flow Cytometry
FN50
APC-eFluor™ 780
CD69
Liquid
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Videos
Documentos