Learn More
CD64, Pacific Blue™, clone: 10.1, Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen CD6428
Detalles adicionales : Peso : 0.00050kg
Descripción
This gene encodes a protein that plays an important role in the immune response. This protein is a high-affinity Fc-gamma receptor. The gene is one of three related gene family members located on chromosome 1.
Our Pacific Blue™ dye is optimally excited by the violet laser and conjugates of this dye are strongly fluorescent even at neutral pH. Pacific Blue™ and Pacific Orange™ dye conjugates can be simultaneously excited at 405 nm and emit at 455 nm and 551 nm, respectively, facilitating two-color analysis.Especificaciones
CD64 | |
Monoclonal | |
Pacific Blue | |
FCGR1A | |
RP11-196G18.2, CD64, CD64A, FCRI, FLJ18345, IGFR1, Fc fragment of IgG, high affinity Ia, receptor for CD64, Fc gamma receptor, Fc-gamma RI, Fc-gamma receptor 1 | |
Mouse | |
Affinity Purified | |
ASR, ASR/IVD | |
2209 | |
Store reagents at 2-8°C. | |
Liquid |
Flow Cytometry | |
10.1 | |
PBS with BSA and 0.1% sodium azide | |
P12314 | |
FCGR1A | |
Human CD64 | |
0.5 mL | |
Primary | |
Human | |
Antibody | |
IgG1 |