missing translation for 'onlineSavingsMsg'
Learn More

CD64 Mouse anti-Mouse, APC-eFluor® 780, Clone: X54-5/7.1, eBioscience™

Mouse Monoclonal Antibody

Marca:  Invitrogen 47-0641-82

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15923294

  • 325.00€ / 100 microgramos


Fecha estimada de envío
20-08-2020
Añadir a la cesta

Descripción

Descripción

Fixation: Samples can be stored in IC Fixation Buffer (Product No. 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (Product No. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 633-647 nm; Emission: 780 nm; Laser: Red Laser CD64 (FCGR1A) encodes a protein that plays an important role in the immune response. CD64 is a high-affinity Fc-gamma receptor and is one of three related gene family members located on chromosome 1. CD64 plays an important role in the immune response and its dysfunction has been studied in cervical adenitis and tetrasomy 21.
Especificaciones

Especificaciones

CD64
Monoclonal
0.2 mg/mL
PBS with 0.09% sodium azide; pH 7.2
P26151
Fc-gamma RI, FcgRI
Mouse
IgG1, kappa
100 μg
4° C, store in dark, DO NOT FREEZE!
Primary
14129
Flow Cytometry
X54-5/7.1
APC-eFluor™ 780
Fcgr1
Liquid
Fcgr1
BALB/c mouse CD64-human IgG Fc fusion protein
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse
Videos
Documentos