Learn More
CD62L, APC, clone: Dreg-56, Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MHCD62L05
Detalles adicionales : Peso : 0.50000kg
Descripción
CD62L (L-selectin) is an adhesion glycoprotein that is constitutively expressed on the cell surface of leukocytes and mediates their homing to inflammatory sites and peripheral lymph nodes by enabling rolling along the venular wall. CD62L is also involved in activation-induced neutrophil aggregation. Activation-dependent CD62L shedding, however, counteracts neutrophil rolling. CD62L has also signaling roles including enhance of chemokine receptor expression. Similarly to CD62P, the major ligand of CD62L is PSGL-1 (P-selectin glycoprotein ligand-1).
Allophycocyanin (APC) is a stable and highly soluble phycobiliprotein that provides maximal absorbance and fluorescence without susceptibility to internal or external fluorescence quenching, thus providing exceptional quantum yields and molar extinction coefficients.Especificaciones
CD62L | |
Monoclonal | |
APC | |
SELL | |
SELL, LAM-1, LAM1, LECAM1, LNHR, LSEL, LYAM1, Leu-8, Lyam-1, PLNHR, TQ1, hLHRc | |
Mouse | |
Purified | |
ASR, ASR/IVD | |
6402 | |
Store at 2 - 8 °C. | |
Liquid |
Flow Cytometry | |
Dreg-56 | |
PBS with 4mg/mL BSA, sucrose and 0.1% sodium azide | |
P14151 | |
SELL | |
Human CD62L | |
0.5 mL | |
Primary | |
Human | |
Antibody | |
IgG1 κ |