missing translation for 'onlineSavingsMsg'
Learn More

CD58/LFA-3 Antibody (085), DyLight 594, Novus Biologicals™

Rabbit Monoclonal Antibody

Marca:  Novus Biologicals NBP2-90096DL594

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18213746

  • 607.98€ / 0.10 ml
Fecha estimada de envío: 14-08-2024
para ver el stock

Descripción

Descripción

CD58/LFA-3 Monoclonal specifically detects CD58/LFA-3 in Human samples. It is validated for Western Blot.
Especificaciones

Especificaciones

CD58/LFA-3
Monoclonal
DyLight 594
50mM Sodium Borate
Rabbit
Protein A purified
RUO
Primary
Human
Purified
Western Blot
85
Western Blot
Ag3, CD58 antigen, CD58 antigen, (lymphocyte function-associated antigen 3), CD58 molecule, FLJ23181, FLJ43722, LFA-3, LFA3ag3, lymphocyte function-associated antigen 3, Surface glycoprotein LFA-3
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD58/LFA-3 (AAH05930.1; Met1-Arg215).
0.1 mL
Cell Biology, Cellular Markers, Immunology
965
Store at 4°C in the dark.
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales