missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD57 Mouse anti-Human, BB515, Clone: NK-1, BD Horizon™
Mouse Monoclonal Antibody
Marca: BD Biosciences 565945
Detalles adicionales : Peso : 0.01000kg
Descripción
CD57 Monoclonal antibody specifically detects CD57 in Human samples. It is validated for Flow cytometry.Especificaciones
CD57 | |
Monoclonal | |
Brilliant Blue 515 (BB515) | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BB515 under optimum conditions and unconjugated antibody was removed. | |
IgM κ |
Flow Cytometry | |
NK-1 | |
Vol. Per test: 5μL | |
B3GAT1; Beta-1,3-glucuronyltransferase 1; HNK1; LEU7; NK-1; GLCATP; GLCUATP | |
25 Tests | |
Cell Biology | |
Human | |
Purified |
For Reseach Use Only.