missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICAM-1 Monoclonal Antibody (YN1/1.7.4), Pacific Blue™, Invitrogen™
Rat Monoclonal Antibody
Marca: Invitrogen A15404
Detalles adicionales : Peso : 0.00280kg
Descripción
ICAM-1 Monoclonal antibody specifically detects ICAM-1 in Mouse, Porcine samples. It is validated for Flow Cytometry.Especificaciones
ICAM-1 | |
Monoclonal | |
0.5 mg/mL | |
PBS with <0.1% sodium azide | |
P13597 | |
ICAM1 | |
Mouse NS-1 cells | |
100 μg | |
Primary | |
Mouse, Porcine | |
Antibody | |
IgG2b κ |
Flow Cytometry | |
YN1/1.7.4 | |
Pacific Blue | |
ICAM1 | |
BB2; CD54; cell surface glycoprotein P3.58; Human rhinovirus receptor; ICAM; Icam1; ICAM-1; ICAM-1; CD54 homolog; Intercellular adhesion molecule 1; intercellular adhesion molecule 1 (CD54), human rhinovirus receptor; intercellular adhesion molecule-1; intercellular adhesion molecule-1 precursor; Ly 47; Ly-47; major group rhinovirus receptor; MALA2; MALA-2; MyD10; P3.58; sCD54; soluble CD54; Surface antigen of activated B cells | |
Rat | |
purified | |
RUO | |
15894, 396750 | |
4° C, store in dark | |
Liquid |