missing translation for 'onlineSavingsMsg'
Learn More

CD5 Mouse anti-Human, Super Bright 780, Clone: UCHT2, eBioscience™

Mouse Monoclonal Antibody

Marca:  Invitrogen 78-0059-41

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15924124

  • 135.00€ / paquete de 25


Fecha de envío estimada
del proveedor 05-07-2019
Añadir a la cesta

Descripción

Descripción

Please refer to the datasheet for Super Bright Staining Buffer for more information. In some experiments, we have observed that compensation values for Super Bright 780-conjugated antibodies are higher in the violet 450/50 channel when using UltraComp eBeads microspheres (Product No. 01-2222-42) as compared to single-color stained cells. In such circumstances, we would recommend setting compensation with cells. We have also observed this in some experiments using AbC Total Antibody Compensation beads (Product No. A10497). Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (Product No. 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (Product No. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 405 nm; Emission: 780 nm; Laser: Violet Laser CD5 is a 67 kDa human T-lymphocyte single-chain transmembrane glycoprotein. CD5 is present on all mature T-lymphocytes, on most of thymocytes and on many T-cell leukemias and lymphomas. CD5 also reacts with a subpopulation of activated B-cells and may act as a receptor in regulating T-cell proliferation. CD5 is found on 95% of thymocytes and 72% of peripheral blood lymphocytes. In lymph nodes, the main reactivity is observed in T cell areas. CD5 is expressed by many T cell leukemia, lymphomas, and activated T cells. Diseases associated with CD5 dysfunction include thymus cancer and Richter’s Syndrome.
Especificaciones

Especificaciones

CD5
Monoclonal
5 μL/Test
PBS with BSA and 0.09% sodium azide; pH 7.2
P06127
CD5 antigen p56-62, T-cell surface glycoprotein CD5, lymphocyte antigen T1/Leu-1
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
UCHT2
Super Bright 780
CD5
Liquid
CD5
IgG1, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
921
Videos