missing translation for 'onlineSavingsMsg'
Learn More

CD45R (B220) Rat, anti-Mouse, PE, Clone: RA3-6B2, STEMCELL Technologies

Rat Monoclonal Antibody

85.12€ - 168.91€

Especificaciones

Antígeno CD45R (B220)
Clon RA3-6B2
Aplicaciones Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
Clasificación Monoclonal
Conjugado PE
Ver más especificaciones

Productos 2
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
17187581
Ver documentos Detalles de la promoción
Stemcell Technologies
100-0420
25 μg
85.12€
25 microgramos
Ver productos alternativos
Ahorre hasta 
»
Fecha estimada de envío: 22-04-2024
para ver el stock
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
17197581
Ver documentos Detalles de la promoción
Stemcell Technologies
100-0421
100 μg
168.91€
100 microgramos
Ver productos alternativos
Ahorre hasta 
»
Fecha estimada de envío: 22-04-2024
para ver el stock
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
Descripción

Descripción

  • The RA3-6B2 antibody reacts with an epitope on the extracellular domain of the exon A‐restricted isoform of mouse CD45
  • CD45 is a member of the protein tyrosine phosphatase family and functions in a number of immunoregulatory processes, including cell activation, growth, differentiation, and oncogenic transformation
  • The large cytoplasmic portion of CD45 contains two tyrosine phosphatase domains (one of which is enzymatically active) that are involved in modulating the function of Src kinases such as Lck and Fyn
  • Several isoforms of CD45 have been identified, the expression of which differs according to cell type and functional status
  • Alternative splicing of three exons encoding the extracellular RA, RB, and RC polypeptide sequences gives rise to up to 8 isoforms with molecular masses in the range of 200 - 220 kDa
  • This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Mouse B Cell Isolation Kit (Catalog #19854)
Especificaciones

Especificaciones

CD45R (B220)
Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
PE
Feline, Human, Mouse
RA3-6B2
Monoclonal
Rat
Primary
Videos
SDS
Documentos

Documentos

Product Certifications