missing translation for 'onlineSavingsMsg'
Learn More

CD45 Rat anti-Mouse, Super Bright 780, Clone: 30-F11, eBioscience™

Rat Monoclonal Antibody

Marca:  Invitrogen 78-0451-82

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15924154

  • 282.00€ / 100 microgramos


Fecha estimada de envío
25-08-2020
Añadir a la cesta

Descripción

Descripción

In some experiments, we have observed that compensation values for Super Bright 780-conjugated antibodies are higher in the violet 450/50 channel when using UltraComp eBeads microspheres (Product No. 01-2222-42) as compared to single-color stained cells. In such circumstances, we would recommend setting compensation with cells. We have also observed this in some experiments using AbC Total Antibody Compensation beads (Product No. A10497). Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (Product No. 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (Product No. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 405 nm; Emission: 780 nm; Laser: Violet Laser Super Bright Polymer Dyes are sold under license from Becton, Dickinson and Company. CD45 (LCA, leukocyte common antigen) is a receptor-type protein tyrosine phosphatase (PTP) ubiquitously expressed in all nucleated hematopoietic cells, comprising approximately 10% of all surface proteins in lymphocytes. CD45 is absent on non-hematopoietic cell lines, normal and maligt, non-hematopoietic tissues. CD45 glycoprotein is crucial in lymphocyte development and antigen signaling, serving as an important regulator of Src-family kinases. CD45 protein exists as multiple isoforms as a result of alternative splicing, differ in their extracellular domains but share identical transmembrane and cytoplasmic domains. CD45RA is an isoform of the CD45 complex and has restricted expression between different subtypes of lymphoid cells. CD45 isoforms differ in their ability to translocate into the glycosphingolipid-enriched membrane domains and their expression depends on cell type and physiological state of the cell. CD45 has been shown to be an essential regulator of T- and B-cell antigen receptor signaling and suppresses JAK kinases to regulate cytokine receptor signaling. CD45 is also important in promoting cell survival by modulating integrin-mediated signal transduction pathway, DNA fragmentation during apoptosis and inhibition or upregulation of various immunological functions.
Especificaciones

Especificaciones

CD45
Monoclonal
0.2 mg/mL
PBS with BSA and 0.09% sodium azide; pH 7.2
P06800
Cluster designation 45; common leukocyte antigen; T220 and B220; HCD45
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse
Flow Cytometry
30-F11
Super Bright 780
Ptprc
Liquid
Ptprc
IgG2b, kappa
100 μg
4° C, store in dark, DO NOT FREEZE!
Primary
19264
Videos
Documentos