Learn More
CD45 Mouse, anti-Human, FITC, Clone: HI30, STEMCELL Technologies
Mouse Monoclonal Antibody
Marca:  Stemcell Technologies 60018FI.1
Detalles adicionales : Peso : 0.00110kg
 Ver más versiones de este producto
Código de producto. 17188491
  • 114.38€ / 25 tests
Fecha estimada de envío: 08-11-2022
para ver el stock
Descripción
Descripción
  • The HI30 antibody reacts with all isoforms of CD45, a type I transmembrane glycoprotein expressed on the surface of most hematopoietic cells except mature erythrocytes, platelets, and plasma cells; expression of CD45 is lost during differentiation of these cell types
  • CD45 is a member of the protein tyrosine phosphatase family and functions in a number of immunoregulatory processes, including cell activation, growth, differentiation, and oncogenic transformation
  • The large cytoplasmic portion of CD45 contains two tyrosine phosphatase domains that are involved in modulating the function of intracellular substrates such as the Src kinases Lck and Fyn
  • This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Human T Cell Enrichment Kit (Catalog #19051), EasySep™ Human CD4+ T Cell Enrichment Kit (Catalog #19052) and EasySep™ Human CD45 Depletion Kit II (Catalog #17898; partial blocking may be observed)
Especificaciones
Especificaciones
CD45
Monoclonal
FITC
25 Tests
Human, Chimpanzee
Granulocytes and Subsets, Hematopoietic Stem and Progenitor Cells, Leukocytes, Lymphocytes, Myeloid Cells
Functional Assay, Immunofluorescence, Immunocytochemistry, Flow Cytometry, CyTOF
HI30
Mouse
Primary
IgG1 κ