missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD44 Rat, anti-Mouse, Alexa Fluor 488, Clone: IM7, STEMCELL Technologies
Rat Monoclonal Antibody
182.25€ - 407.70€
Especificaciones
Antígeno | CD44 |
---|---|
Clon | IM7 |
Aplicaciones | Immunohistochemistry, ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence |
Clasificación | Monoclonal |
Conjugado | Alexa Fluor 488 |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
17198811
|
Stemcell Technologies
60068AD.1 |
25 μg |
182.25€
25 microgramos |
Fecha estimada de envío: 10-05-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
17188811
|
Stemcell Technologies
60068AD |
100 μg |
407.70€
100 microgramos |
Fecha estimada de envío: 10-05-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
Descripción
- The IM7 antibody reacts with CD44 (Ly-24), an ∼80 - 95 kDa type 1 transmembrane glycoprotein involved in cell-cell and cell-matrix interactions
- CD44 is expressed on the surface of many cells, including leukocytes and hepatocytes, as well as endothelial, epithelial, and mesenchymal cells
- Expression levels increase upon activation of T and B cells, and memory cells exhibit a CD44 high phenotype
- CD44 binds many ligands, including hyaluronic acid, collagen, fibronectin, growth factors, and metalloproteinases, thus modulating processes such as lymphocyte activation, recirculation and homing, leukocyte rolling and aggregation, hematopoiesis, and tumor metastasis
- This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Mouse CD4+ T Cell Isolation Kit (Catalog #19852), EasySep™ Mouse CD4+CD62L+ T Cell Isolation Kit (Catalog #18765), and EasySep™ Human Naive CD4+ T Cell Isolation Kit (Catalog #19555)
Especificaciones
CD44 | |
Immunohistochemistry, ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
Alexa Fluor 488 | |
Baboon, Cynomolgus, Rhesus Monkey, Chimpanzee, Human | |
Primary |
IM7 | |
Monoclonal | |
Rat | |
IgG2b κ |