missing translation for 'onlineSavingsMsg'
Learn More

CD44 Antibody (065), Alexa Fluor™ 532, Novus Biologicals™

Rabbit Monoclonal Antibody

Marca:  Novus Biologicals NBP2-90077AF532

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18213617

  • 607.98€ / 0.10 ml
Fecha estimada de envío: 19-08-2024
para ver el stock

Descripción

Descripción

CD44 Monoclonal specifically detects CD44 in Human samples. It is validated for Western Blot, ELISA.
Especificaciones

Especificaciones

CD44
Monoclonal
Alexa Fluor 532
50mM Sodium Borate
Rabbit
Protein A purified
RUO
Primary
Human
Purified
Western Blot, ELISA
65
Western Blot, ELISA
CD44 antigen, CD44 molecule (Indian blood group), CD44R, CDw44, cell surface glycoprotein CD44, chondroitin sulfate proteoglycan 8, CSPG8, ECMR-III, Epican, Extracellular matrix receptor III, GP90 lymphocyte homing/adhesion receptor, HCELL, hematopoietic cell E- and L-selectin ligand, Heparan sulfate proteoglycan, Hermes antigen, homing function and Indian blood group system, HUTCH-I, Hyaluronate receptor, IN, LHR, MC56, MDU2CD44 antigen (homing function and Indian blood group system), MDU3CDW44, MIC4MGC10468, Pgp1, PGP-1, PGP-I, Phagocytic glycoprotein 1, Phagocytic glycoprotein I
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD44 (P16070-1; Met 1-Pro 220).
0.1 mL
Angiogenesis, Apoptosis, Cancer, Cell Biology, Core ESC Like Genes, Extracellular Matrix, Hematopoietic Stem Cell Markers, Immunology, Mesenchymal Stem Cell Markers, Oncogenes, Phospho Specific, Protein Kinase, Signal Transduction, Stem Cell Markers
960
Store at 4°C in the dark.
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales