Learn More
CD42b/GPIb alpha Antibody (HIP1), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Marca:  Bio-Techne NB500-511UV
Detalles adicionales : Peso : 0.00970kg
Código de producto. 18216080
  • 491.35€ / 0.10 ml
Fecha estimada de envío: 19-08-2024
para ver el stock
Descripción
Descripción
CD42b/GPIb alpha Monoclonal specifically detects CD42b/GPIb alpha in Human, Mouse, Primate samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.
Especificaciones
Especificaciones
CD42b/GPIb alpha
Monoclonal
DyLight 350
Antigen CD42b-alpha, BSS, CD42b, CD42b antigen, CD42b-alpha, glycoprotein Ib (platelet), alpha polypeptide, Glycoprotein Ibalpha, GP1B, GP-Ib alpha, GPIbA, GPIb-alpha, MGC34595, platelet glycoprotein Ib alpha chain, platelet membrane glycoprotein 1b-alpha subunit
Mouse
Affinity Purified
RUO
Primary
The antibody HIP1 reacts with CD42b (GPIb alpha), a 135-145 kDa membrane glycoprotein expressed on platelets and megakaryocytes. CD42b and CD42c (GPIb beta) are composed in a disulfide linked heterodimer (CD42b/c; 160 kDa); CD42b/c forms a noncovalent complex with CD42a and CD42d. HLDA IV; WS Code P 40
Store at 4C in the dark.
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
HIP1
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen
GP1BA
Peripheral blood mononuclear cells of a patient suffering with CLL.
0.1 mL
Immunology
2811
Human, Mouse, Primate
IgG1