Learn More
CD4 Rat, anti-Mouse, PerCP-Cyanine5.5, Clone: RM4-5, STEMCELL Technologies
Rat Monoclonal Antibody
Marca:  Stemcell Technologies 60017PS.1
Detalles adicionales : Peso : 0.00110kg
 Ver más versiones de este producto
Código de producto. 17198481
  • 186.20€ / 25 microgramos
Fecha estimada de envío: 14-11-2022
para ver el stock
Descripción
Descripción
  • The RM4-5 antibody reacts with mouse CD4, an ∼55 kDa single-chain type 1 transmembrane glycoprotein and member of the immunoglobulin (Ig) superfamily; CD4 contains four extracellular Ig-like domains
  • CD4 is expressed at relatively high levels by most thymocytes and a subpopulation of T cells (T helper/inducer cells), and at lower levels on dendritic cells
  • CD4 binds to a non-polymorphic region of MHC II and acts as a co-receptor to the T cell receptor (TCR) in MHC II-restricted antigen recognition by enhancing the avidity of the association between the TCR and MHC II-antigen complex
  • Binding of the RM4-5 antibody to CD4 inhibits ligand binding in vitro; binding of the RM4-5 antibody can be blocked by the clone GK1.5 antibody
  • This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Mouse CD4+ T Cell Isolation Kit (Catalog #19852) and EasySep™ Mouse CD25 Regulatory T Cell Positive Selection Kit (Catalog #18782)
Especificaciones
Especificaciones
CD4
Monoclonal
PerCP-Cyanine5.5
25 μg
Mouse
T Cells, T Cells, CD4+, T Cells, Regulatory
Immunofluorescence, Immunocytochemistry, Flow Cytometry, Western Blot, Immunohistochemistry
RM4-5
Rat
Primary
IgG2a κ