Learn More
CD4 Mouse, anti-Human, APC, Clone: OKT4, STEMCELL Technologies
Mouse Monoclonal Antibody
Marca:  Stemcell Technologies 60016AZ.1
Detalles adicionales : Peso : 0.00110kg
 Ver más versiones de este producto
Código de producto. 17188461
  • 144.97€ / 25 tests
Fecha estimada de envío: 08-11-2022
para ver el stock
Descripción
Descripción
  • The OKT4 antibody reacts with CD4, an ∼59 kDa single-chain type 1 transmembrane glycoprotein and member of the immunoglobulin (Ig) superfamily; CD4 contains four extracellular Ig-like domains (D1 - D4)
  • The epitope for OKT4 has been localized to the D3 domain of the protein, which has a structure resembling an Ig variable domain
  • CD4 is expressed at high levels by most thymocytes and a subpopulation of T cells (T-helper cells), and at lower levels by peripheral blood monocytes and macrophages
  • Moreover, CD4 is a receptor for human immunodeficiency virus (HIV)
  • Binding of the OKT4 antibody to CD4 does not block HIV binding
  • This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Direct Human CD4+ T Cell Isolation Kit (Catalog #19662), EasySep™ Human CD4+ T Cell Enrichment Kit (Catalog #19052), EasySep™ Human CD3 Positive Selection Kit II (Catalog #17851) and EasySep™ Human CD4 Positive Selection Kit II (Catalog #17852)
Especificaciones
Especificaciones
CD4
Monoclonal
APC
25 Tests
Human, Chimpanzee, Sooty Mangabey, Rhesus Monkey, Cynomolgus
T Cells, T Cells, CD4+, T Cells, Regulatory
Immunoprecipitation, Functional Assay, Immunofluorescence, Flow Cytometry, Immunohistochemistry
OKT4
Mouse
Primary
IgG2b κ