missing translation for 'onlineSavingsMsg'
Learn More

CD39, Super Bright 600, Clone: eBioA1 (A1), Monoclonal, eBioscience™

Mouse Monoclonal Antibody

Marca:  Invitrogen 63-0399-41

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15923504

  • 135.00€ / paquete de 25


Fecha de envío estimada
del proveedor 02-04-2019
Añadir a la cesta

Descripción

Descripción

It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Super Bright 600 is a tandem dye that can be excited with the violet laser line (405 nm) and emits at 600 nm. We recommend using a 610/20 bandpass filter. Please make sure that your instrument is capable of detecting this fluorochrome. When using two or more Super Bright dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Staining Buffer (cat. SB-4400) to minimize any non-specific polymer interactions. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 2-8°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorochrome performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 405 nm; Emission: 600 nm; Laser: Violet Laser CD39 cell surface antigen is a 70-100kD molecule expressed on peripheral blood B cells, monocytes, T cell clones, and also weakly expressed on granulocytes. CD39 has intrinsic ecto-ATPase activity. Expression of CD39 is induced on T cells and increased on B cells as a late activation antigen.
Especificación

Especificación

CD39
Monoclonal
5 μL/Test
PBS with BSA and 0.09% sodium azide; pH 7.2
P49961
ENTPD1
IgG1, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
953
Flow Cytometry
eBioA1 (A1)
Super Bright 600
ENTPD1
Liquid
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Videos