Learn More
CD39 Monoclonal Antibody (eBioA1 (A1)), Super Bright™ 600, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
en la oferta
Especificaciones
Antígeno | CD39 |
---|---|
Clon | eBioA1 (A1) |
Concentración | 5 μL/Test |
Aplicaciones | Flow Cytometry |
Clasificación | Monoclonal |
Descripción
Description: The eBioA1 monoclonal antibody reacts with human CD39 also known as ectonucleoside triphosphate diphosphohydrolase 1 (ENTPD1) or NTPDase. CD39 is an integral membrane protein with two transmembrane domains and exists as a homotetramer. It is the most prominent ectoenzyme of the immune system. The function of CD39 is to effectively remove toxic extracellular ATP by converting it to ADP or AMP. CD39 is thought to work together with CD73 to hydrolyze ATP and has been well characterized on Langerhans cells. Expression of CD39 was originally identified on activated lymphocytes. Expression is also found on a subset of T cells, B cells and dendritic cells as well as weak staining on monocytes and granulocytes. Recently, CD39 and CD73 have been found on regulatory T cells (Treg). Expression of CD39 on Treg may facilitate their entry into inflamed areas where high levels of ATP are present. Expression of CD39 on Foxp3+CD4+ cells ranges from 25-45%. Applications Reported: This eBioA1 (A1) antibody has been reported for use in flow cytometric analysis. Applications Tested: This eBioA1 (A1) antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cels. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.
CD39 Monoclonal antibody specifically detects CD39 in Human samples. It is validated for Flow Cytometry.Especificaciones
CD39 | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
P49961 | |
ENTPD1 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
ENTPD1 |
eBioA1 (A1) | |
Flow Cytometry | |
Super Bright 600 | |
Mouse | |
Human | |
953 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |