Learn More
CD38 Mouse, anti-Human, FITC, Clone: HIT2, STEMCELL Technologies
Mouse Monoclonal Antibody
Marca:  Stemcell Technologies 60014FI
Detalles adicionales : Peso : 0.00110kg
 Ver más versiones de este producto
Código de producto. 17188441
  • 341.55€ / 100 tests
Fecha estimada de envío: 10-05-2024
para ver el stock
Descripción
Descripción
  • The HIT2 antibody reacts with CD38, an ∼45 kDa type II transmembrane glycoprotein expressed by many cell types, especially leukocytes
  • CD38 is an ectoenzyme (ADP-ribosyl hydrolase) with both cyclase and hydrolase activities, whereby it mediates lymphocyte activation, adhesion and metabolism
  • CD31 is a known ligand for the receptor
  • In humans, CD38 is expressed by the majority of hematopoietic cells at levels which vary according to the differentiation and activation status of the cells
  • It is highly expressed on activated T and B cells and plasma cells, and is also found at high levels in several tissues, including brain, kidney, liver, ovary, pancreas, placenta, testis, and some types of cancerous tissue
  • This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Human CD138 Positive Selection Kit II (Catalog #1877) and EasySep™ Human Whole Blood and Bone Marrow CD138 Positive Selection Kit II (Catalog #17887)
Especificaciones
Especificaciones
CD38
Monoclonal
FITC
100 Tests
Chimpanzee, Human
B Cells, Hematopoietic Stem and Progenitor Cells, Plasma
Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
HIT2
Mouse
Primary
IgG1 κ