missing translation for 'onlineSavingsMsg'
Learn More

CD370 (Clec9A) Rat anti-Mouse, BB515, Clone: 10B4, BD Horizon™

Rat Monoclonal Antibody

Marca:  BD Biosciences 565320

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15824239

  • 343.00€ / cada uno


Fecha estimada de envío
07-02-2020
Añadir a la cesta

Descripción

Descripción

CD370 (Clec9A) Monoclonal antibody specifically detects CD370 (Clec9A) in Mouse samples. It is validated for Flow cytometry.
Especificaciones

Especificaciones

CD370 (Clec9A)
10B4 (also known as 24/04-10B4)
BB515
Purified
Rat
Rat WI, also known as Wistar (outbred) IgG2a, κ
RUO
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BB515 under optimum conditions and unconjugated antibody was removed.
Monoclonal
Flow Cytometry
0.2mg/mL
Aqueous buffered solution containing ≤0.09% sodium azide.
CD370; CLC9A; Clec9a; DNGR-1; C-type lectin domain family 9 member A
Mouse Clec9A Peptide
50μg
Cell Biology
Primary
Murine
Videos
Documentos

For Research Use Only.