missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD32 Rat, Qfluor 630, Clone: 2.4G2, enQuireBio™
Rat Monoclonal Antibody
Marca: EnquireBio QAB87-QF630-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
CD32 | |
Monoclonal | |
0.5mg/mL | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
FCGR2b | |
Protein A/G Purified | |
RUO | |
14131 | |
IgG2b |
Flow Cytometry | |
2.4G2 | |
Qfluor 630 | |
XP_006496722 | |
Rat | |
500 μg | |
Primary | |
Store at 2-4°C. |